accessibility menu, dialog, popup

UserName

CH3SBOHHO

CAS RN 145349-76-4

IUPAC Name: [4-(ethylsulfanyl)phenyl]boronic acid
Synonyms: 4-(ethylsulfanyl)phenylboronic acid
Molecular Weight (g/mol): 182.04
Molecular Formula: C8H11BO2S
InChi Key: LMUASBOUSAMURX-UHFFFAOYSA-N
SMILES: CCSC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results