accessibility menu, dialog, popup

UserName

BONCH3H3CFHOOH

CAS RN 1451391-88-0

IUPAC Name: [3-(dimethylcarbamoyl)-2-fluorophenyl]boronic acid
Synonyms: 3-(dimethylcarbamoyl)-2-fluorophenylboronic acid
Molecular Weight (g/mol): 211
Molecular Formula: C9H11BFNO3
InChi Key: ICJWWRWLFPUVJI-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=CC(B(O)O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results