accessibility menu, dialog, popup

UserName

CAS RN 1449145-31-6

BSOONCH3CH3HOOH

CAS RN 1449145-31-6

IUPAC Name: [3-(dipropylsulfamoyl)phenyl]boronic acid
Synonyms: 3-(dipropylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 285.17
Molecular Formula: C12H20BNO4S
InChi Key: NUYSZOMJDYKRCE-UHFFFAOYSA-N
SMILES: CCCN(CCC)S(=O)(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results