accessibility menu, dialog, popup

UserName

CAS RN 1449131-72-9

BFOHNNHOOH

CAS RN 1449131-72-9

IUPAC Name: {4-fluoro-3-[(pyridin-2-yl)carbamoyl]phenyl}boronic acid
Synonyms: 4-fluoro-3-[(pyridin-2-yl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 260.03
Molecular Formula: C12H10BFN2O3
InChi Key: DJNCGHBHBJUZKA-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(C(=O)NC2=CC=CC=N2)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results