CAS RN 144371-88-0
CAS RN 144371-88-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
FR 901379 (>90%), TRC
CAS: 144371-88-0 Synonym: 1-[(4R,5R)-4,5-Dihydroxy-N2-(1-oxohexadecyl)-L-ornithine]-4-[(4S)-4-hydroxy-4-[4-hydroxy-3-(sulfooxy)phenyl]-L-threonine]pneumocandin A0∣(2α,3β,4β)-(4R,5R)-4,5-Dihydroxy-N2-(1-oxohexadecyl)-L-ornithyl-L-threonyl-trans-4-hydroxy-L-prolyl-(S)-4-hydroxy-4-[4-hydroxy-3-(sulfooxy)phenyl]-L-threonyl-3-hydroxy-L-glutaminyl-3-hydroxy-4-methyl-L-Proline Cyclic (6→1)-Peptide∣1H-Dipyrrolo[2,1-c:2',1'-l][1,4,7,10,13,16]hexaazacycloheneicosine Cyclic Peptide Deriv.