accessibility menu, dialog, popup

UserName

CAS RN 14435-88-2

CH3NH3C

CAS RN 14435-88-2

IUPAC Name: 2,6-bis(4-methylphenyl)pyridine
Synonyms: 2,6-bis(4-methylphenyl)pyridine
Molecular Weight (g/mol): 259.35
Molecular Formula: C19H17N
InChi Key: BLRYHCTZTSRBNB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=CC(=N1)C1=CC=C(C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results