accessibility menu, dialog, popup

UserName

CAS RN 1435806-19-1

ClOFFFOHONH2

CAS RN 1435806-19-1

IUPAC Name: 2-amino-3-[4-chloro-2-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 2-amino-3-[4-chloro-2-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 283.63
Molecular Formula: C10H9ClF3NO3
InChi Key: JHAAXZPMBGSSFH-UHFFFAOYNA-N
SMILES: NC(CC1=C(OC(F)(F)F)C=C(Cl)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)