CAS RN 142977-64-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2-difluoro-3-phenylpropanoic acid, TRC
CAS: 142977-64-8 Molecular Formula: C9H8O2F2 Molecular Weight (g/mol): 186.16 SMILES: OC(=O)C(F)(F)Cc1ccccc1