CAS RN 142691-70-1
CAS RN 142691-70-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1'-Bis(dichlorophosphino)ferrocene, 98%, Thermo Scientific Chemicals
CAS: 142691-70-1 Molecular Formula: C10H8Cl4FeP2 Molecular Weight (g/mol): 387.77 MDL Number: MFCD01630854 IUPAC Name: 1,1'-Bis(dichlorophosphino)ferrocene SMILES: [Fe].ClP(Cl)c1cccc1.ClP(Cl)c1cccc1