CAS RN 1423-10-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Fluorobenzene-d5, TRC
CAS: 1423-10-5 Molecular Formula: C6 2H5 F Molecular Weight (g/mol): 101.13 Synonym: Fluorobenzene-d5,Fluorobenzene D5 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-fluorobenzene SMILES: [2H]c1c([2H])c([2H])c(F)c([2H])c1[2H]
2
Fluorobenzene-d5, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.