CAS RN 1421373-62-7
CAS RN 1421373-62-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mutant EGFR inhibitor, MedChemExpress
MedChemExpress Mutant EGFR inhibitor is a potent and selective mutant EGFR inhibitor extracted from patent WO 2013014448 A1; inhibits EGFRL858R, EGFRExon 19 deletion and EGFRT790M.