CAS RN 14197-60-5
CAS RN 14197-60-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
20(S)-Ginsenoside Rg3, MedChemExpress
MedChemExpress 20(S)-Ginsenoside Rg3 is the main component of Red ginseng. Ginsenoside Rg3 inhibits Na+ and hKv1.4 channel with IC50s of 32.2±4.5 and 32.6±2.2 μM, respectively. 20(S)-Ginsenoside Rg3 also inhibits Aβ levels, NF-κB activity, and COX-2 expression.