accessibility menu, dialog, popup

UserName

CAS RN 14173-30-9

HClHONHO

CAS RN 14173-30-9

IUPAC Name: hydrogen 2-(hydroxymethyl)pyridin-3-ol chloride
Synonyms: hydrogen 2-(hydroxymethyl)pyridin-3-ol chloride
Molecular Weight (g/mol): 161.59
Molecular Formula: C6H8ClNO2
InChi Key: SKVRYQUMKJQZFN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OCC1=NC=CC=C1O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results