accessibility menu, dialog, popup

UserName

OHOOHNOHO

CAS RN 138-60-3

IUPAC Name: 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid
Synonyms: 4-oxo-1H-pyridine-2,6-dicarboxylic acid
Molecular Weight (g/mol): 183.12
Molecular Formula: C7H5NO5
InChi Key: XTLJJHGQACAZMS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C=C(N1)C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)