accessibility menu, dialog, popup

UserName

CAS RN 13767-89-0

SOOOHHOCaNa22

CAS RN 13767-89-0

IUPAC Name: calcium disodium bis(sulfuric acid)
Synonyms: calcium disodium bis(sulfuric acid)
Molecular Weight (g/mol): 282.2
Molecular Formula: CaH4Na2O8S2
InChi Key: GYZGFUUDAQXRBT-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Ca++].OS(O)(=O)=O.OS(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results