accessibility menu, dialog, popup

UserName

H3COFOOH

CAS RN 137654-21-8

IUPAC Name: 2-fluoro-6-methoxybenzoic acid
Synonyms: 2-fluoro-6-methoxybenzoic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: XLNQGZLCGVLNMF-UHFFFAOYSA-N
SMILES: COC1=C(C(O)=O)C(F)=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results