accessibility menu, dialog, popup

UserName

CAS RN 1374508-59-4

BOOH3CCH3H3CH3CFONH2

CAS RN 1374508-59-4

IUPAC Name: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 265.09
Molecular Formula: C13H17BFNO3
InChi Key: ZETAUVYXFMYTIN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(C(N)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results