accessibility menu, dialog, popup

UserName

CAS RN 1373921-08-4

CH3HOFFFF

CAS RN 1373921-08-4

IUPAC Name: [4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]methanol
Synonyms: [4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 208.16
Molecular Formula: C9H8F4O
InChi Key: XVYKBFIHKYRUPA-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC(CO)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results