accessibility menu, dialog, popup

UserName

CH3OOHFFFF

CAS RN 1373921-03-9

IUPAC Name: [2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]methanol
Synonyms: [2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 224.16
Molecular Formula: C9H8F4O2
InChi Key: ZWHWVTLVMXSITQ-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C(CO)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results