accessibility menu, dialog, popup

UserName

CAS RN 1373920-94-5

H3COFNF

CAS RN 1373920-94-5

IUPAC Name: 4-butoxy-2,6-difluorobenzonitrile
Synonyms: 4-butoxy-2,6-difluorobenzonitrile
Molecular Weight (g/mol): 211.21
Molecular Formula: C11H11F2NO
InChi Key: DRMAQHPUVUFLAE-UHFFFAOYSA-N
SMILES: CCCCOC1=CC(F)=C(C#N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results