accessibility menu, dialog, popup

UserName

FFNCl

CAS RN 1373920-84-3

IUPAC Name: 2-(6-chloro-2,3-difluorophenyl)acetonitrile
Synonyms: 2-(6-chloro-2,3-difluorophenyl)acetonitrile
Molecular Weight (g/mol): 187.57
Molecular Formula: C8H4ClF2N
InChi Key: DZNGMYTZQJRVAF-UHFFFAOYSA-N
SMILES: FC1=C(F)C(CC#N)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results