accessibility menu, dialog, popup

UserName

CAS RN 1373920-73-0

CH3OOHFFFF

CAS RN 1373920-73-0

IUPAC Name: 3-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 3-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 250.19
Molecular Formula: C11H10F4O2
InChi Key: ROIPUVGZXPJVDC-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC(CCC(O)=O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results