CAS RN 137218-25-8
CAS RN 137218-25-8
IUPAC Name:
2-[2-(trifluoromethoxy)phenyl]acetonitrile
Synonyms:
2-[2-(trifluoromethoxy)phenyl]acetonitrile
Molecular Weight (g/mol):
201.15
Molecular Formula:
C9H6F3NO
InChi Key:
OIKWCWVMUBCXJM-UHFFFAOYSA-N
SMILES:
FC(F)(F)OC1=CC=CC=C1CC#N
1
–
1
of
1
results
1
2-(Trifluoromethoxy)phenylacetonitrile, 98%, Thermo Scientific™
CAS: 137218-25-8 Molecular Formula: C9H6F3NO Molecular Weight (g/mol): 201.15 MDL Number: MFCD00236326 InChI Key: OIKWCWVMUBCXJM-UHFFFAOYSA-N Synonym: 2-trifluoromethoxy phenylacetonitrile,2-triflnoromethoxybenzyl cyanide,2-2-trifluoromethoxy phenyl acetonitrile,2-trifluoromethoxybenzyl cyanide,1 2-trifluoromethoxy phenyl acetonitrile,2-trifluoromethoxy benzyl cyanide,2-trifluoromethoxybenzylcyanide,2-trifluoromethoxy-phenyl-acetonitrile,benzeneacetonitrile, 2-trifluoromethoxy PubChem CID: 2777321 IUPAC Name: 2-[2-(trifluoromethoxy)phenyl]acetonitrile SMILES: FC(F)(F)OC1=CC=CC=C1CC#N