CAS RN 1370256-78-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SKLB4771, MedChemExpress
MedChemExpress SKLB4771 is a novel potent and selective Flt3 inhibitor with IC50 of 10 nM; against FLT3-ITD-expressing MV4-11 cells with IC50 of 6 nM.