accessibility menu, dialog, popup

UserName

H3CH3CCH3NOH(E)

CAS RN 13559-66-5

IUPAC Name: N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine
Synonyms: N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine
Molecular Weight (g/mol): 167.25
Molecular Formula: C10H17NO
InChi Key: OVFDEGGJFJECAT-DHZHZOJONA-N
SMILES: CC1(C)C2CCC1(C)\C(C2)=N\O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results