accessibility menu, dialog, popup

UserName

SNNHNCl

CAS RN 134796-34-2

IUPAC Name: 5-{[(4-chlorophenyl)methyl]sulfanyl}-1H-1,2,4-triazole
Synonyms: 3-{[(4-chlorophenyl)methyl]sulfanyl}-2H-1,2,4-triazole
Molecular Weight (g/mol): 225.69
Molecular Formula: C9H8ClN3S
InChi Key: IPYLPWUQKVUWGY-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CSC2=NC=NN2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results