CAS RN 13446-19-0
IUPAC Name:
magnesium(2+) hexahydrate diperchlorate
Synonyms:
magnesium(2+) hexahydrate diperchlorate
Molecular Weight (g/mol):
331.29
Molecular Formula:
Cl2H12MgO14
InChi Key:
MGBDNCBRZIMSDZ-UHFFFAOYSA-L
SMILES:
O.O.O.O.O.O.[Mg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
1
–
1
of
1
results
1
Magnesium perchlorate hexahydrate, 99% (metals basis)
CAS: 13446-19-0 Molecular Formula: Cl2H12MgO14 Molecular Weight (g/mol): 331.287 MDL Number: MFCD00149778 InChI Key: MGBDNCBRZIMSDZ-UHFFFAOYSA-L Synonym: magnesium perchlorate hexahydrate,acmc-20akl9,2clo4.mg.6h2o,ksc491a3f,magnesium 2+ ion hexahydrate diperchlorate,magnesium 2+ hexahydrate diperchlorate PubChem CID: 16211969 IUPAC Name: magnesium;diperchlorate;hexahydrate SMILES: O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]