accessibility menu, dialog, popup

UserName

CH3OSOHOCl

CAS RN 133659-14-0

IUPAC Name: 5-chloro-4-methoxythiophene-3-carboxylic acid
Synonyms: 5-chloro-4-methoxythiophene-3-carboxylic acid
Molecular Weight (g/mol): 192.61
Molecular Formula: C6H5ClO3S
InChi Key: CEOFOUVVEMMYDW-UHFFFAOYSA-N
SMILES: COC1=C(Cl)SC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results