CAS RN 1330180-72-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Glycidyl Myristate-d5, TRC
CAS: 1330180-72-7 Molecular Formula: C17H27D5O3 Molecular Weight (g/mol): 289.46 Synonym: Tetradecanoic Acid 2-Oxiranylmethyl-d5 Ester,Glycidyl-d5 Tetradecanoate,Myristic Acid Glycidyl-d5 Ester,NSC 404224-d5; IUPAC Name: [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] tetradecanoate SMILES: O=C(OC([2H])([2H])C1([2H])OC1([2H])[2H])CCCCCCCCCCCCC