accessibility menu, dialog, popup

UserName

CAS RN 13221-86-8

NH2HNOOHHO

CAS RN 13221-86-8

IUPAC Name: 2,4-dihydroxybenzohydrazide
Synonyms: 2,4-dihydroxybenzohydrazide
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: GOPUHTXVZGIOHN-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(O)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results