CAS RN 13189-98-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
S-(3-Hydroxypropyl)-L-cysteine, TRC
CAS: 13189-98-5 Molecular Formula: C6 H13 N O3 S Molecular Weight (g/mol): 179.24 Synonym: Fudosteine IUPAC Name: (2R)-2-amino-3-(3-hydroxypropylsulfanyl)propanoic acid SMILES: N[C@@H](CSCCCO)C(=O)O
2
Fudosteine, MedChemExpress
MedChemExpress Fudosteine is a cysteine derivative and a mucoactive agent. Fudosteine inhibits MUC5AC mucin hypersecretion by reducing MUC5AC gene expression.