accessibility menu, dialog, popup

UserName

CAS RN 1315281-51-6

BOOH3CCH3H3CH3CN

CAS RN 1315281-51-6

IUPAC Name: 9-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}-9H-carbazole
Synonyms: 9-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}carbazole
Molecular Weight (g/mol): 383.3
Molecular Formula: C25H26BNO2
InChi Key: AKZHQKVQCWCVBW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CN2C3=C(C=CC=C3)C3=C2C=CC=C3)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results