accessibility menu, dialog, popup

UserName

CAS RN 131123-76-7

OHOOClClNH

CAS RN 131123-76-7

IUPAC Name: 5,7-dichloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
Synonyms: 5,7-dichlorokynurenic acid
Molecular Weight (g/mol): 258.05
Molecular Formula: C10H5Cl2NO3
InChi Key: BGKFPRIGXAVYNX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C2=C(Cl)C=C(Cl)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results