accessibility menu, dialog, popup

UserName

CAS RN 13110-37-7

H3COONH2

CAS RN 13110-37-7

IUPAC Name: pentyl 4-aminobenzoate
Synonyms: pentyl 4-aminobenzoate
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: VKYWCHMXHQTCJQ-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results