CAS RN 130812-47-4
IUPAC Name:
barium(2+) bis(2-ethoxyethan-1-olate)
Synonyms:
barium(2+) bis(2-ethoxyethanolate)
Molecular Weight (g/mol):
315.56
Molecular Formula:
C8H18BaO4
InChi Key:
WCXRGQPXPCQKSJ-UHFFFAOYSA-N
SMILES:
[Ba++].CCOCC[O-].CCOCC[O-]
1
–
1
of
1
results
1
Barium 1-methoxy-2-propoxide, 25% w/w in 1-methoxy-2-propanol, Thermo Scientific™
CAS: 130812-47-4 Molecular Formula: C8H18BaO4 Molecular Weight (g/mol): 315.555 MDL Number: MFCD00078021 InChI Key: WCXRGQPXPCQKSJ-UHFFFAOYSA-N Synonym: bis 2-ethoxyethoxy barium,barium methoxypropoxide w/w in methoxypropanol,barium bis 2-ethoxyethan-1-olate,barium 2+ ion bis 2-ethoxyethanolate PubChem CID: 14619883 IUPAC Name: barium(2+);2-ethoxyethanolate SMILES: CCOCC[O-].CCOCC[O-].[Ba+2]