CAS RN 130793-28-1
CAS RN 130793-28-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Formyl-L-leucine (1S)-1-[(2R,3S)-3-Carboxy-2-hydroxynonyl]dodecyl Ester, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.