accessibility menu, dialog, popup

UserName

CAS RN 130714-47-5

CH3O(R)(R)OH(R)OHO(R)NNHNNN

CAS RN 130714-47-5

IUPAC Name: (2R,3R,4R,5R)-5-[6-(cyclohexylamino)-9H-purin-9-yl]-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
Synonyms: (2R,3R,4R,5R)-5-[6-(cyclohexylamino)purin-9-yl]-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
Molecular Weight (g/mol): 363.42
Molecular Formula: C17H25N5O4
InChi Key: JAKAFSGZUXCHLF-LSCFUAHRSA-N
SMILES: CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(NC3CCCCC3)N=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results