accessibility menu, dialog, popup

UserName

RhSbFFFFFF

CAS RN 130296-28-5

IUPAC Name: bis(cycloocta-1,5-diene) hexafluorostibanuide rhodium
Synonyms: bis(1,5-cyclooctadiene) hexafluorostibanuide rhodium
Molecular Weight (g/mol): 555.02
Molecular Formula: C16H24F6RhSb
InChi Key: ZSPLJWNAXJRVRK-UHFFFAOYSA-H
SMILES: [Rh].F[Sb-](F)(F)(F)(F)F.C1CC=CCCC=C1.C1CC=CCCC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)