CAS RN 1281816-04-3
CAS RN 1281816-04-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lys-SMCC-DM1, MedChemExpress
MedChemExpress Lys-SMCC-DM1 (Lys-Nε-MCC-DM1) is a linker-payload component that has the potential to inhibit tubulin polymerization.Lys-SMCC-DM1 is the active metabolite of T-DM1. T-DM1 is a human epidermal growth factor receptor 2 (HER2)-targeting ADC with a tubulin polymerization inhibitor DM1.