CAS RN 127779-20-8
CAS RN 127779-20-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Saquinavir, MedChemExpress
MedChemExpress Saquinavir(Ro 31-8959) is an HIV Protease inhibitor used in antiretroviral therapy. Saquinavir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 1.36 μM.