CAS RN 1276197-42-2
CAS RN 1276197-42-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Terbufos-d10 (O,O-diethyl-d10), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Terbufos-d10 (O,O-diethyl-d10), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
Terbufos-d10 (O,O-diethyl-d10), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Terbufos-d10 (O,O-diethyl-d10), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.