CAS RN 127337-60-4
CAS RN 127337-60-4
IUPAC Name:
2-(chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride
Synonyms:
2-(chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride
Molecular Weight (g/mol):
276.08
Molecular Formula:
C9H10Cl2F3NO
InChi Key:
CMZBQUWICURDCD-UHFFFAOYSA-N
SMILES:
Cl.CC1=C(OCC(F)(F)F)C=CN=C1CCl
1
–
1
of
1
results
1
2-Chloromethyl-3-methyl-4-(2,2,2-trifluoro-ethoxy)pyridine Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.