CAS RN 1272-44-2
CAS RN 1272-44-2
IUPAC Name:
(2-benzoylcyclopenta-2,4-dien-1-yl)(cyclopenta-2,4-dien-1-yl)iron
Synonyms:
(2-benzoylcyclopenta-2,4-dien-1-yl)(cyclopenta-2,4-dien-1-yl)iron
Molecular Weight (g/mol):
290.14
Molecular Formula:
C17H14FeO
InChi Key:
JJHHJZCYDPLHIL-UHFFFAOYNA-N
SMILES:
O=C(C1=CC=CC1[Fe]C1C=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Benzoylferrocene, 98%, Thermo Scientific Chemicals
CAS: 1272-44-2 Molecular Formula: C17H14FeO Molecular Weight (g/mol): 290.14 MDL Number: MFCD00001431 InChI Key: JJHHJZCYDPLHIL-UHFFFAOYNA-N Synonym: Ferrocenyl phenyl ketone IUPAC Name: Benzoylferrocene SMILES: [Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1