CAS RN 1272-21-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
352.54
Molecular Formula:
C15H15Gd
InChi Key:
YAYGFOZNMGYPRE-UHFFFAOYSA-N
SMILES:
[Gd+3].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[CH-]1C=CC=C1
1
–
1
of
1
results
1
Tris(cyclopentadienyl)gadolinium(III), 99.9% (REO), Thermo Scientific™
CAS: 1272-21-5 Molecular Formula: C15H15Gd Molecular Weight (g/mol): 352.535 MDL Number: MFCD00145476 InChI Key: WPVCUPCREAIUBK-UHFFFAOYSA-N Synonym: tris cyclopentadienyl gadolinium-gd reo PubChem CID: 73411059 IUPAC Name: cyclopenta-1,3-diene;gadolinium(3+) SMILES: C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Gd+3]