CAS RN 1271-66-5
CAS RN 1271-66-5
IUPAC Name:
bis(cyclopenta-1,3-diene) bis(methane) titanium
Synonyms:
bis(cyclopentadiene) bis(methane) titanium
Molecular Weight (g/mol):
212.16
Molecular Formula:
C12H20Ti
InChi Key:
CRKLPNOLYBORCP-UHFFFAOYSA-N
SMILES:
C.C.[Ti].C1C=CC=C1.C1C=CC=C1
1
–
1
of
1
results
1
Promotions Available
Bis(cyclopentadienyl)dimethyltitanium, 5 wt.% in toluene, AcroSeal™