CAS RN 1270014-40-8
CAS RN 1270014-40-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
BTK IN-1, MedChemExpress
MedChemExpress BTK IN-1 (SNS062 analog) is a potent BTK inhibitor, with an IC50 of <100 nM.