CAS RN 1265917-14-3
CAS RN 1265917-14-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S1RA hydrochloride, MedChemExpress
MedChemExpress S1RA hydrochloride (E-52862 hydrochloride) is a potent and selective sigma-1 receptor(σ1R, Ki=17 nM) antagonist, showed good selectivity against σ2R (Ki > 1000 nM).