CAS RN 1263774-59-9
CAS RN 1263774-59-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Delgocitinib, MedChemExpress
MedChemExpress Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.