CAS RN 1263273-14-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1R,2S)-VU0155041, MedChemExpress
MedChemExpress (1R,2S)-VU0155041, Cis regioisomer of VU0155041, is a partial mGluR4 agonist with an EC50 of 2.35 μM.