accessibility menu, dialog, popup

UserName

CAS RN 1259994-92-7

CH3FFFOOHH2N

CAS RN 1259994-92-7

IUPAC Name: 2-amino-3-[2-methyl-5-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 2-amino-3-[2-methyl-5-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 247.22
Molecular Formula: C11H12F3NO2
InChi Key: UQIIXNIENIPXSV-UHFFFAOYNA-N
SMILES: CC1=C(CC(N)C(O)=O)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)